missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCCIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16043-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene product was isolated on the basis of its interaction with BRCA2 and p21 proteins. It is an evolutionarily conserved nuclear protein with multiple interacting domains. The N-terminal half shares moderate homology with regions of calmodulin and M-calpain, suggesting that it may also bind calcium. Functional studies indicate that this protein may be an important cofactor for BRCA2 in tumor suppression, and a modulator of CDK2 kinase activity via p21. This protein has also been implicated in the regulation of BRCA2 and RAD51 nuclear focus formation, double-strand break-induced homologous recombination, and cell cycle progression. Multiple transcript variants encoding different isoforms have been described for this gene.Spécification
BCCIP | |
Polyclonal | |
Unconjugated | |
BCCIP | |
BCCIP, Protein TOK 1, TOK 1, TOK1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
361666, 56647, 66165 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunocytochemistry, Immunofluorescence | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CWI3, Q9P287 | |
Bccip | |
BCCIP Fusion Protein Ag8934 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |