missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCL2L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16026-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Using a PCR-based strategy, an additional protein with life-promoting activity, designated Bcl-w, has been identified. The protein is highly conserved between mouse and human and is encoded by a gene located near the TCRalpha gene on chromosome 14. Bcl-w is expressed in myeloid cell lines but not in T and B lymphocytes, and can be found in a wide range of tissues. An alternative splicing event in exon 4 results in two transcripts. The first, Bcl-w, encodes a protein of 193 amino acids, and the second, Bcl-w/rox, encodes a protein 333 amino acids in length. The "rox" portion of Bcl-w/rox shows a striking 66% amino acid sequence identity with the Drosophila rox2 protein; however, the Bcl-w/rox transcript may be expressed at very low levels.Spécification
BCL2L2 | |
Polyclonal | |
Unconjugated | |
BCL2L2 | |
Apoptosis regulator Bcl W, BCL2L2, KIAA0271 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12050, 599, 60434 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P70345, Q92843 | |
BCL2L2 | |
BCL2L2 Fusion Protein Ag8863 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |