missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDKRB1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26672-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Bradykinin, a 9 aa peptide, is generated in pathophysiologic conditions such as inflammation, trauma, burns, shock, and allergy. Two types of G-protein coupled receptors have been found which bind bradykinin and mediate responses to these pathophysiologic conditions. The protein encoded by this gene is one of these receptors and is synthesized de novo following tissue injury. Receptor binding leads to an increase in the cytosolic calcium ion concentration, ultimately resulting in chronic and acute inflammatory responses.Spécification
BDKRB1 | |
Polyclonal | |
Unconjugated | |
BDKRB1 | |
B1 bradykinin receptor, B1BKR, B1R, BDKRB1, BK 1 receptor, BKB1R, BKR1, BRADYB1, bradykinin receptor B1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
623 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P46663 | |
BDKRB1 | |
BDKRB1 Fusion Protein Ag24745 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |