missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biglycan Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16409-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a small cellular or pericellular matrix proteoglycan that is closely related in structure to two other small proteoglycans, decorin and fibromodulin. The encoded protein and decorin are thought to be the result of a gene duplication. Decorin contains one attached glycosaminoglycan chain, while this protein probably contains two chains. For this reason, this protein is called biglycan. This protein plays a role in assembly of collagen fibrils and muscle regeneration. It interacts with several proteins involved in muscular dystrophy, including alpha-dystroglycan, alpha- and gamma-sarcoglycan and collagen VI, and it is critical for the assembly of the dystrophin-associated protein complex.Spécification
Biglycan | |
Polyclonal | |
Unconjugated | |
BGN | |
A6NLG9, BGN, biglycan, Bone/cartilage proteoglycan I, DSPG1, PG S1, PGI, SLRR1A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12111, 25181, 397396, 633 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P21810, P28653, P47853, Q9GKQ6 | |
BGN | |
Biglycan Fusion Protein Ag0683 | |
150 μL | |
Primary | |
Human, Mouse, Porcine, Rat | |
Antibody | |
IgG |