Learn More
Bim Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 22037-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16831585
  • 399.00€ / 150µL
Expédition estimée: 13-05-2024
Description
The Bim antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Bim. This antibody recognizes human, mouse antigen. The Bim antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Spécification
Bim
Polyclonal
Unconjugated
150 μL
10018
IgG
Western Blot, Immunohistochemistry, ELISA
300 μg/mL
Rabbit
Primary
Human, Mouse