Learn More
Bim Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22037-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The Bim antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Bim. This antibody recognizes human, mouse antigen. The Bim antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
Bim | |
Polyclonal | |
Unconjugated | |
150 μL | |
10018 | |
IgG |
Western Blot, Immunohistochemistry, ELISA | |
300 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |