missing translation for 'onlineSavingsMsg'
Learn More

BPIL1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13461-2-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16831694

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a member of the lipid transfer/lipopolysaccharide binding protein gene family. It is highly expressed in hypertrophic tonsils. This gene and three other members of the LT/LBP gene family form a cluster on the long arm of chromosome 20.
Spécification

Spécification

BPIL1
Polyclonal
Unconjugated
BPIFB2
BPIL1, C20orf184, LPLUNC2, RYSR
Rabbit
Antigen Affinity Chromatography
RUO
80341
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8N4F0
BPIFB2
BPIL1 Fusion Protein Ag4051
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions