missing translation for 'onlineSavingsMsg'
Learn More

BSAP/PAX5 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 26709-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16835095

  • 325.38€ / 150µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is specific to PAX5. It will not cross react with other PAX members.

PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.
Spécification

Spécification

BSAP/PAX5
Polyclonal
Unconjugated
PAX5
BSAP, BSAP,PAX5, paired box 5, Paired box protein Pax 5, PAX5
Rabbit
Antigen Affinity Chromatography
RUO
5079
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q02548
PAX5
BSAP,PAX5 Fusion Protein Ag25147
150 μL
Primary
Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions