missing translation for 'onlineSavingsMsg'
Learn More

BTN2A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 16482-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836124

  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Butyrophilin is the major protein associated with fat droplets in the milk. This gene is a member of the BTN2 subfamily of genes, which encode proteins belonging to the butyrophilin protein family. The gene is located in a cluster on chromosome 6, consisting of seven genes belonging to the expanding B7/butyrophilin-like group, a subset of the immunoglobulin gene superfamily. The encoded protein is a type I receptor glycoprotein involved in lipid, fatty-acid and sterol metabolism. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Spécification

Spécification

BTN2A2
Polyclonal
Unconjugated
BTN2A2
BT2.2, BTF2, BTN2A2
Rabbit
Antigen Affinity Chromatography
RUO
10385, 238555, 306957
-20°C
Liquid
Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A4QPC6, Q8WVV5
BTN2A2
BTN2A2 Fusion Protein Ag9603
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions