missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C-Peptide Antibody (CII-29), Novus Biologicals™
Mouse Monoclonal Antibody
436.00€
Spécification
| Antigène | C-Peptide |
|---|---|
| Clone | CII-29 |
| Dilution | ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence, Immunoassay, Sandwich ELISA 1:100-1:2000 |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunoassay |
| Classification | Monoclonal |
Description
C-Peptide Monoclonal antibody specifically detects C-Peptide in Mouse, Rat samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunoassay.Spécification
| C-Peptide | |
| ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence, Immunoassay, Sandwich ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| 3630 | |
| Reacts with rat c-peptide II, mouse c-peptide II, mouse c-peptide I, rat c-peptide I and rat proinsulin | |
| Primary | |
| Store at 4C. Do not freeze. |
| CII-29 | |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunoassay | |
| Unconjugated | |
| Rat | |
| Diabetes Research | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| INS | |
| IgG1 | |
| Protein A purified | |
| Reacts with rat c-peptide II, mouse c-peptide II, mouse c-peptide I, rat c-peptide I and rat proinsulin |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit