missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C14orf2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14704-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
MP68, also known as PLPM or C14orf2 (chromosome 14 open reading frame 2), is a 58 amino acid mitochondrial protein that belongs to the small mitochondrial proteolipid family. The gene encoding MP68 maps to human chromosome 14, which houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer′s disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder alpha1-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.Spécification
C14orf2 | |
Polyclonal | |
Unconjugated | |
MP68 | |
C14orf2, MP68, PLPM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9556 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P56378 | |
MP68 | |
C14orf2 Fusion Protein Ag6386 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |