Learn More
C15orf55 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28639-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The C15orf55 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human C15orf55. This antibody recognizes human, mouse antigen. The C15orf55 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Spécification
| C15orf55 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 256646 | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 500 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |