missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf14 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20808-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
C16orf14 | |
Polyclonal | |
Unconjugated | |
MCRIP2 | |
C16orf14, c349E10.1, FAM195A, Protein FAM195A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
68241, 84331 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9BUT9, Q9CQB2 | |
Mcrip2 | |
C16orf14 Fusion Protein Ag14754 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |