missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C17orf46 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24629-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
C17orf46, also named as SPATA32 and TEX34, mainly expressed in Testis. It interacts with syntaxin-1 and ACTB. It′s about 42-52 kDa in our detection.Spécification
C17orf46 | |
Polyclonal | |
Unconjugated | |
SPATA32 | |
C17orf46 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
124783 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96LK8 | |
SPATA32 | |
C17orf46 Fusion Protein Ag18239 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |