Learn More
C17orf49 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 27114-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16835405
  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
Description
Component of chromatin complexes such as the MLL1/MLL and NURF complexes.
Spécification
Spécification
C17orf49
Polyclonal
Unconjugated
BAP18
C17orf49
Rabbit
Antigen Affinity Chromatography
RUO
104457, 124944
-20°C
Liquid
Western Blot, Immunoprecipitation
0.53 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IXM2, Q9DCT6
BAP18
C17orf49 Fusion Protein Ag25882
150 μL
Primary
Mouse, Human