missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C19orf44 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21503-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The function of this protein remains unknown.Spécification
C19orf44 | |
Polyclonal | |
Unconjugated | |
C19orf44 | |
C19orf44 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
290615, 84167 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9H6X5 | |
C19orf44, RGD1311847 | |
C19orf44 Fusion Protein Ag15960 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |