missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf163 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25361-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Required for assembly of mitochondrial respiratory chain complex I and complex IV.Spécification
C1orf163 | |
Polyclonal | |
Unconjugated | |
COA7 | |
C1orf163, COA7, RESA1, Sel1 repeat-containing protein 1, SELRC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
65260, 69893 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q921H9, Q96BR5 | |
Coa7 | |
C1orf163 Fusion Protein Ag19734 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |