missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3orf31 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 28137-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
C3orf31 may be involved in the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.Spécification
C3orf31 | |
Polyclonal | |
Unconjugated | |
TAMM41 | |
C3orf31, DKFZp434E0519 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
132001 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96BW9 | |
TAMM41 | |
C3orf31 Fusion Protein Ag26826 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |