missing translation for 'onlineSavingsMsg'
Learn More

C6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 17239-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16897084

  • 349.00€ / 150µL
Expédition estimée: 21-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

C6 is a component of complement cascade. It is part of the membrane attack complex which can insert into the cell membrane and cause cell to lyse. People with C6 deficiency are prone to bacterial infection. Two transcript variants encoding the same protein have been found for this gene.
Spécification

Spécification

C6
Polyclonal
Unconjugated
C6
C6, complement component 6, Complement component C6
Rabbit
Antigen Affinity Chromatography
RUO
12274, 24237, 729
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P13671, Q811M5
C6
C6 Fusion Protein Ag11083
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions