missing translation for 'onlineSavingsMsg'
Learn More

C6orf182 Rabbit anti-Human, Mouse, Non-human primate, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 24957-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16833485

  • 325.38€ / 150µL
Expédition estimée: 10-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

C6orf182 also named as Centrosomal protein of 57 kDa related protein is a 460 amino-acid protein, which belongs to translokin family. C6orf182 as centrosomal protein may be required for microtubule attachment to centrosomes and localizes in the cytoplasm. The function of C6orf182 need to be further studied.
Spécification

Spécification

C6orf182
Polyclonal
Unconjugated
CEP57L1
C6orf182, Centrosomal protein 57kDa-like protein 1, Cep57 related protein, CEP57L1, Cep57R
Rabbit
Antigen Affinity Chromatography
RUO
103268, 285753
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry
0.31 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8IYX8, Q8VDS7
CEP57L1
C6orf182 Fusion Protein Ag21054
150 μL
Primary
Non-human Primate, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions