Learn More
Epredia™ Lab Vision™ CA19-9/Sialyl Lewis a (GI Tumor Marker) Ab-3, Mouse Monoclonal Antibody
Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ CA19-9/Sialyl Lewisa (GI Tumor Marker) Ab-3, Mouse Monoclonal Antibody.
Marque: Epredia™ MS-1811-R7
Description
Host Species: Mouse
Clone: 116-NS-19-9
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Sialic acid-containing residue of the monosialoganglioside
Immunogen: Human colon carcinoma cell line SW1116
Positive Control: Colon CA
Cellular Localization: Cell membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Spécification
| CA19-9/Sialyl Lewissuperscriptasuperscript (GI Tumor Marker) Ab-3 | |
| Monoclonal | |
| Unconjugated | |
| Human colon carcinoma cell line SW1116 | |
| RUO | |
| Primary | |
| Prediluted |
| Immunohistochemistry (Paraffin) | |
| 116-NS-19-9 | |
| Mouse | |
| 7 mL | |
| Oncogenesis | |
| Human | |
| IgG2a κ |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.