missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 11071-1-AP-20UL
Weitere Details : Poids : 0.00010kg
Beschreibung
Carbonic anhydrase (CA) is an enzyme that assists rapid interconversion of carbon dioxide and water into carbonic acid, protons, and bicarbonate ions. It is abundant in all mammalian tissues. There are many genes that are inducible by hypoxia, via HIF-1 alpha. CA IX is one of the most inducible genes because of its stability and location within the membrane. Carbonic anhydrases have a widespread role in regulating pH in normal tissues, by regulating hydrogen ion (H+) flux. The pH is important in cell death under hypoxia, thus a blockade of CA IX results in increased cell death under hypoxia. Therefore, CA IX has become a reliable histochemical marker of hypoxia.Spezifikation
CA9 | |
Polyclonal | |
Unconjugated | |
CA9 | |
CA IX, CA9, CAIX, Carbonate dehydratase IX, Carbonic anhydrase 9, carbonic anhydrase IX, G250, Membrane antigen MN, MN, P54/58N, pMW1, RCC associated antigen G250 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230099, 313495, 768 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation, Flow Cytometry | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q16790, Q8VHB5 | |
CA9, Car9 | |
CA9 Fusion Protein Ag1540 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |