missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-13 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | Cadherin-13 |
---|---|
Concentration | 0.22 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16830434
|
Proteintech
12618-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 13-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16820434
|
Proteintech
12618-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 13-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The protein encoded by this gene has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3′ trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.Spécification
Cadherin-13 | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
P55290, Q9WTR5 | |
1012, 12554 | |
Cadherin-13 Fusion Protein Ag3295 | |
Primary | |
-20°C | |
CDH13 |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Cadherin 13, Cadherin-13, CDH13, CDHH, H cadherin, H-cadherin, Heart cadherin, P105, T cad, T cadherin, T-cadherin, Truncated cadherin | |
CDH13 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |