Learn More
CAMK1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 24312-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16832795
  • 325.38€ / 150µL
Expédition estimée: 28-09-2022
pour voir le stock disponible 
Description
Description
The CAMK1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human CAMK1. This antibody recognizes human antigen. The CAMK1 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Spécification
Spécification
CAMK1
Polyclonal
Unconjugated
150 μL
8536
IgG
ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
300 μg/mL