missing translation for 'onlineSavingsMsg'
Learn More

Carboxypeptidase A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15836-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16835204

  • 121.38€ / 20µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is specific to CPA1.

Three different forms of human pancreatic procarboxypeptidase A have been isolated. This gene encodes a monomeric pancreatic exopeptidase involved in zymogen inhibition.
Spécification

Spécification

Carboxypeptidase A1
Polyclonal
Unconjugated
CPA1
Carboxypeptidase A1, CPA, CPA1
Rabbit
Antigen Affinity Chromatography
RUO
109697, 1357, 24269
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P00731, P15085, Q7TPZ8
Cpa1
Carboxypeptidase A1 Fusion Protein Ag8586
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions