missing translation for 'onlineSavingsMsg'
Learn More

CCDC85B Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 18282-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16828354

  • 325.38€ / 150µL
Expédition estimée: 03-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

DIPA functions as a transcriptional repressor. It may inhibit the activity of CTNNB1 in a TP53-dependent manner and thus regulate cell growth. It may function in adipocyte differentiation, negatively regulating mitotic clonal expansion.
Spécification

Spécification

CCDC85B
Polyclonal
Unconjugated
CCDC85B
CCDC85B, Delta interacting protein A, DIPA
Rabbit
Antigen Affinity Chromatography
RUO
11007, 240514
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15834, Q6PDY0
CCDC85B
CCDC85B Fusion Protein Ag13129
150 μL
Primary
Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions