missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC9 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21104-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Coiled-coil domain-containing protein 9 (CCDC9) contains one coiled-coil domain.Spécification
CCDC9 | |
Polyclonal | |
Unconjugated | |
CCDC9 | |
CCDC9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
243846, 26093 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VC31, Q9Y3X0 | |
Ccdc9 | |
CCDC9 Fusion Protein Ag15374 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |