Learn More
CCR4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque:  Novus Biologicals NBP1-86584
Code nomenclature Nacres: NA.46
449.65 EUR valable jusqu'au 2025-12-16
MEILLEUR PRIX promo! Utilisez le code promo "24090" pour bénéficier de cette offre.
Code produit. 18776263
  • 573.00€ / 0.10mL
Description
CCR4 Polyclonal specifically detects CCR4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Spécification
Spécification
Polyclonal
CCR4
RUO
Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20-1:50
This antibody was developed against Recombinant Protein corresponding to amino acids:LQIYSADTPSSSYTQSTMDHDLHDAL
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
For Research Use Only