Learn More
CCR7 (CD197) Mouse anti-Human , APC, Clone: 2-L1-A, BD Biosciences™
Mouse Monoclonal Antibody
160.50€ - 343.47€
Spécification
| Antigène | CCR7 (CD197) |
|---|---|
| Clone | 2-L1-A |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugué | APC |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
17207594
|
BD Biosciences
566763 |
25 Tests |
161.00€ 160.50€ / 25 tests Économisez 0.50€ 0.31% de réduction |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
17297584
|
BD Biosciences
566762 |
100 Tests |
344.00€ 343.47€ / 100 tests Économisez 0.53€ 0.15% de réduction |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow CytometrySpécification
| CCR7 (CD197) | |
| Flow Cytometry | |
| APC | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human CCR7 Transfected Cell Line | |
| Primary |
| 2-L1-A | |
| Monoclonal | |
| Mouse | |
| Human | |
| BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to APC under optimum conditions, and unconjugated antibody and free APC were removed. |