missing translation for 'onlineSavingsMsg'
Learn More

CCR7 (CD197) Mouse anti-Human , PE-CF594, Clone: 2-L1-A, BD Biosciences™

Mouse Monoclonal Antibody

Marque:  BD Biosciences 566768

Informations supplémentaires : Poids : 0.00060kg

 Afficher plus de versions de ce produit

Code produit. 17257594

  • 406.00€ / 100 tests
Expédition estimée: 19-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CCR7 (CD197) Monoclonal antibody specifically detects CCR7 (CD197) in Human samples. It is validated for Flow Cytometry
Spécification

Spécification

CCR7 (CD197)
Monoclonal
PE-CF594
BLR2; CC chemokine receptor 7; CMKBR7; EBI1; EVI1; Epstein-Barr virus induced gene 1; MIP-3 beta receptor
Human CCR7 Transfected Cell Line
RUO
Human
Mouse BALB/c IgG1 κ
Flow Cytometry
2-L1-A
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ PE-CF594 under optimum conditions, and unconjugated antibody and free PE-CF594 were removed.
Vidéos
FDS
Documents
Certificats
Promotions

Promotions