Learn More
CD117 (c-Kit) Monoclonal Antibody (ACK2), Super Bright™ 436, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marque: Affymetrix eBioscience 62-1172-80
Informations supplémentaires : Poids : 0.01000kg
Description
Description: The ACK2 monoclonal antibody reacts with mouse CD117, also known as c-Kit receptor, Steel factor receptor and stem cell factor receptor. A member of the tyrosine kinase receptor family, this 145 kDa molecule is expressed by a majority of hematopoietic progenitor cells characterized in the mouse bone marrow as a small subset of cells positive for Sca-1 and Thy1 (Thy1^lo) and negative for lineage markers. The interaction of the mouse c-kit receptor and steel factor promotes the proliferation and differentiation of hematopoietic progenitor cells. CD117 is also expressed by mast cells and plays a role in signaling and activation of these cells. ACK2 has been reported to be a blocking antibody. Applications Reported: This ACK2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ACK2 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 436 can be excited with the violet laser line (405 nm) and emits at 436 nm. We recommend using a 450/50 bandpass filter, or equivalent.
CD117 (c-Kit) Monoclonal antibody specifically detects CD117 (c-Kit) in Mouse samples. It is validated for Flow Cytometry.Spécification
CD117 (c-Kit) | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
P05532 | |
KIT | |
Affinity chromatography | |
RUO | |
16590 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
ACK2 | |
Super Bright 436 | |
KIT | |
belly-spot; Bs; CD117; ckit; C-Kit; c-KIT gene1; c-kit protein; c-kit proto-oncogene protein; c-kit receptor; c-kit receptor tyrosine kinase; dominant spotting; Dominant white spotting; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; KIT; kit oncogene; KIT proto-oncogene receptor tyrosine kinase; KIT proto-oncogene, receptor tyrosine kinase; mast cell growth factor receptor; mast/stem cell growth factor receptor; mast/stem cell growth factor receptor Kit; MGF; p145 c-kit; PBT; Piebald trait protein; protein kinase; proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; receptor tyrosine kinase c-kit; receptor tyrosine kinase type III; SCFR; SCO1; SCO5; Sl; soluble KIT variant 1; SOW3; spotted sterile male; Ssm; Steel Factor Receptor; Tr-kit; tyrosine kinase receptor; tyrosine kinase receptor c-KIT; tyrosine-protein kinase Kit; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein; W | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b κ |