Learn More
CD137 Ligand (4-1BB Ligand) Monoclonal Antibody (4H3), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
394.00€
Spécification
Antigène | CD137 Ligand (4-1BB Ligand) |
---|---|
Clone | 4H3 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15825373
|
Affymetrix eBioscience
25-5906-42 |
100 Tests |
394.00€
100 pièces |
Expédition estimée: 15-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
This 4H3 monoclonal antibody recognizes human CD137 ligand, also known as 4-1BB ligand, TNFSF9, or CDw137L. Applications Reported: This 4H3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 4H3 antibody has been pre-diluted and tested by flow cytometric analysis of stimulated normal human peripheral blood cells. This may be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (Product # 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (Product # 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD137 Ligand (4-1BB Ligand) Monoclonal antibody specifically detects CD137 Ligand (4-1BB Ligand) in Human samples. It is validated for Flow Cytometry.Spécification
CD137 Ligand (4-1BB Ligand) | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
P41273 | |
TNFSF9 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
TNFSF9 |
4H3 | |
Flow Cytometry | |
PE-Cyanine7 | |
Mouse | |
Human | |
8744 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |