missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD138/Syndecan-1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10593-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene is a transmembrane (type I) heparan sulfate proteoglycan and is a member of the syndecan proteoglycan family. The syndecans mediate cell binding, cell signaling, and cytoskeletal organization and syndecan receptors are required for internalization of the HIV-1 tat protein. The syndecan-1 protein functions as an integral membrane protein and participates in cell proliferation, cell migration and cell-matrix interactions via its receptor for extracellular matrix proteins. Altered syndecan-1 expression has been detected in several different tumor types. While several transcript variants may exist for this gene, the full-length natures of only two have been described to date. These two represent the major variants of this gene and encode the same protein.Spécification
| CD138/Syndecan-1 | |
| Polyclonal | |
| Unconjugated | |
| SDC1 | |
| CD138, SDC, SDC1, SYND1, syndecan, syndecan 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20969, 25216, 6382 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P18827, P18828, P26260 | |
| SDC1 | |
| CD138/Syndecan-1 Fusion Protein Ag0823 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu