missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Mouse anti-Human, Mouse, Clone: 2C1D9, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60253-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.Spécification
CD14 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08571, P10810 | |
CD14 | |
CD14 Fusion Protein Ag10693 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2C1D9 | |
Unconjugated | |
CD14 | |
CD14, CD14 molecule | |
Mouse | |
Protein G | |
RUO | |
12475, 929 | |
-20°C | |
Liquid |