missing translation for 'onlineSavingsMsg'
Learn More

CD15 Mouse anti-Human , BV421, Clone: HI98 (also known as HIM1), BD Biosciences™

Mouse Monoclonal Antibody

Marque:  BD Biosciences 567009

Informations supplémentaires : Poids : 0.00020kg

 Afficher plus de versions de ce produit

Code produit. 17267794

  • 289.00€ / 25 tests
Expédition estimée: 02-01-2023
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CD15 Monoclonal antibody specifically detects CD15 in Human samples. It is validated for Flow Cytometry
Spécification

Spécification

CD15
Monoclonal
Brilliant Violet 421
Lewis X; Le-X; X-Hapten; SSEA-1; 3-FAL
Human Mononuclear Cells
RUO
Human
Mouse IgM κ
Flow Cytometry
HI98 (also known as HIM1)
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions