missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio CD152 / CTLA-4 Mouse, PE, Clone: BNI3, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio QAB59-PE-25Tests
Informations supplémentaires : Poids : 0.01000kg
Spécification
CD152 / CTLA-4 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CTLA4 | |
Protein A/G Purified | |
RUO | |
1493 | |
IgG2a κ |
Flow Cytometry | |
BNI3 | |
PE | |
NP_001032720 | |
Mouse | |
25 Tests | |
Primary | |
Store at 2-4°C. |