missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD155/PVR Antibody (19C12), Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-26355-100ul
459.00 EUR valable jusqu'au 2025-12-16
MEILLEUR PRIX promo! Utilisez le code promo "24090" pour bénéficier de cette offre.
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Beskrivelse
CD155/PVR Monoclonal antibody specifically detects CD155/PVR in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Tekniske data
| CD155/PVR | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| 19C12 | |
| Western Blot 1:500-1:2000, ELISA, Immunohistochemistry 1:50-1:200 | |
| CD155, CD155 antigen, HVED, NECL5, NECL-5, nectin-like 5, Nectin-like protein 5, poliovirus receptor, PVSFLJ25946, Tage4 | |
| A synthesized peptide derived from Human CD155/PVR [UniProt P15151] | |
| 100 μL | |
| Virology Bacteria and Parasites | |
| 5817 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion