Learn More
EnquireBio CD161 Mouse, PerCP-Cy5.5, Clone: PK136, enQuireBio™
Mouse Monoclonal Antibody
Marque:  EnquireBio QAB78-PCP55-100ug
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15982478
  • 328.00€ / 100µg
Expédition estimée: 24-07-2023
Spécification
CD161
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Klrb1c
Protein A/G Purified
RUO
17059
IgG2a κ
Flow Cytometry
PK136
PerCP-Cyanine5.5
NP_001153376
Mouse
100 μg
Primary
Store at 2-4°C.