missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD171 Monoclonal Antibody (eBio5G3 (5G3)), Super Bright™ 436, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
153.00€ - 378.00€
Spécification
Antigène | CD171 |
---|---|
Clone | eBio5G3 (5G3) |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16361234
|
Affymetrix eBioscience
62-1719-41 |
25 Tests |
153.00€
25 pièces |
Expédition estimée: 14-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16371234
|
Affymetrix eBioscience
62-1719-42 |
100 Tests |
378.00€
100 pièces |
Expédition estimée: 14-06-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Mouse Monoclonal Antibody
L1CAM/CD171 is an axonal glycoprotein belonging to the immunoglobulin supergene family. The ectodomain, consisting of several immunoglobulin-like domains and fibronectin-like repeats (type III), is linked via a single transmembrane sequence to a conserved cytoplasmic domain. This cell adhesion molecule plays an important role in nervous system development, including neuronal migration and differentiation. Mutations in the gene cause three X-linked neurological syndromes known by the acronym CRASH (corpus callosum hypoplasia, retardation, aphasia, spastic paraplegia and hydrocephalus). Alternative splicing of a neuron-specific exon is thought to be functionally relevant.Spécification
CD171 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
antigen identified by monoclonal antibody R1; CAML1; CD 171; CD171; CD171 molecule; HSAS; HSAS1; Hyd; L1; L1 cell adhesion molecule; L1CAM; MASA; MIC5; N-CAM L1; NCAML1; N-CAML1; N-CAM-L1; NCAM-L1; Nerve-growth factor-inducible large external glycoprotein; neural cell adhesion molecule L1; NILE; S10; sCD171; sL1 CAM; sL1CAM; soluble CD 171; soluble CD171; soluble L1 CAM; soluble L1CAM; SPG1 | |
L1CAM | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
L1CAM |
eBio5G3 (5G3) | |
Flow Cytometry | |
Super Bright 436 | |
Mouse | |
Human | |
P32004 | |
3897 | |
IgG2a | |
Affinity chromatography | |
Antibody |