missing translation for 'onlineSavingsMsg'
Learn More

CD19 Mouse anti-Human, NovaFluor™ Blue 530, Clone: HIB19, Invitrogen™

Mouse Monoclonal Antibody

Marque:  Invitrogen H004T03B02

Informations supplémentaires : Poids : 0.02000kg

 Afficher plus de versions de ce produit

Code produit. 17256466

  • 350.00€ / 100 pièces
Expédition estimée: 20-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The HIB19 monoclonal antibody reacts with human CD19, a 95 kDa transmembrane glycoprotein. CD19 is expressed by B cells during all stages of development excluding the terminally differentiated plasma cells. Follicular dendritic cells also express CD19. Together CD21, CD81, Leu13, MHC class II, and CD19 form a multimolecular complex that associates with BCR. Signaling through CD19 induces tyrosine phosphorylation, calcium flux and proliferation of B cells. The SJ25C1 antibody and the HIB19 monoclonal antibody recognize overlapping epitopes.
Applications Reported: This HIB19 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This HIB19 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 4 μL (0.6 μg) per test. A test is defined as the amount ( μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
When using more than one NovaFluor dye, CellBlox Monocyte and Macrophage Blocking Buffer may be used to block non-specific binding of dyes to monocytes.
Excitation: 509 nm; Emission: 530 nm; Laser: 488 nm (Blue) Laser

CD19 Monoclonal antibody specifically detects CD19 in Human samples. It is validated for Flow Cytometry
Spécification

Spécification

CD19
Monoclonal
4 μL/Test
PBS with no preservative; pH 7.2
P15391
Mouse
100 Tests
Primary
Human
Antibody
IgG1 κ
Flow Cytometry
HIB19
NovaFluor Blue 530
Cd19
AW495831; B4; B-lymphocyte antigen CD19; B-lymphocyte surface antigen B4; Cd19; CD19 antigen; CD19 molecule; CVID3; differentiation antigen CD19; Leu-12; T-cell surface antigen Leu-12
Affinity chromatography
RUO
930
4° C, store in dark, DO NOT FREEZE!
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions