missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Mouse, PerCP-Cy5.5, Clone: HIB19, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio QAB30-PCP55-25Tests
Informations supplémentaires : Poids : 0.01000kg
Spécification
CD19 | |
Monoclonal | |
5μL/Test | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
CD19 | |
Protein A/G Purified | |
RUO | |
930 |
Flow Cytometry | |
HIB19 | |
PerCP-Cyanine5.5 | |
NP_001171569 | |
Mouse | |
25 Tests | |
Primary | |
IgG1 κ |