Learn More
CD20 Monoclonal Antibody (HI47), Pacific Orange™, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MHCD2030
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00050kg
Description
Our Pacific Orange™ dye is optimally excited by the violet laser, and it is recommended that a 575, 585 or 600 nm band pass filter be used for optimal detection. Pacific Blue™ and Pacific Orange™ dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm and 551 nm, respectively, facilitating two-color analysis.
CD20 Monoclonal antibody specifically detects CD20 in Human samples. It is validated for Flow Cytometry.Spécification
CD20 | |
Monoclonal | |
Pacific Orange | |
Ms4a1 | |
AA960661; APY; ATOPY; B1; B-cell differentiation antigen Ly-44; B-lymphocyte antigen CD20; B-lymphocyte cell-surface antigen B1; B-lymphocyte surface antigen B1; Bp35; Cd20; CD20 antigen; CD20 cell surface protein; CD20 receptor; CVID5; EGK_06167; Fc epsilon receptor I beta chain; Fc Fragment of IgE high affinity I receptor for beta polypeptide; FCER1B; High affinity immunoglobulin epsilon receptor subunit beta; IgE Fc receptor subunit beta; IGEL; IGER; IGHER; LEU16; LEU-16; Leukocyte surface antigen Leu-16; Ly44; Ly-44; Lymphocyte antigen 44; membrane spanning 4-domains A1; membrane-spanning 4-domains subfamily A member 1; membrane-spanning 4-domains, subfamily A, member 1; membrane-spanning 4-domains, subfamily A, member 2; MGC3969; MS4A1; MS4A2; S7 | |
Mouse | |
Affinity chromatography | |
ASR | |
931 | |
4° C, store in dark | |
Liquid |
Flow Cytometry | |
HI47 | |
PBS with BSA and 0.1% sodium azide | |
P11836 | |
Ms4a1 | |
CD20 antigen | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG3 |