Learn More
CD200 Mouse anti-Human, PE, Clone: MRC OX-104, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 552475
Informations supplémentaires : Poids : 0.02000kg
Description
The MRC OX-104 monoclonal antibody specifically binds to CD200. CD200 is a 40-45 kDa type 1 transmembrane glycoprotein and is also referred to as OX2. CD200 is a member of the immunoglobulin superfamily of proteins. It contains two Ig domains, a single transmembrane region and a short cytoplasmic tail. CD200 is expressed on resting and PMA-activated T- and B-cells, but not on NK cells, monocytes, granulocytes, or platelets. It has been found on a subset of CD34+ progenitor cells. Interaction of CD200 with its receptor on macrophages induces a downregulation of macrophage activity in a variety of tissues, suggesting a regulatory function.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Spécification
CD200 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human CD200 Recombinant Protein | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
MRC OX-104 | |
OX-2; MOX1; MOX2 ; My033 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.