Learn More
CD204 Monoclonal Antibody (M204PA), PE, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Marque: Affymetrix eBioscience 12-2046-82
Informations supplémentaires : Poids : 0.01000kg
Beschreibung
This M204PA antibody recognizes mouse CD204 also known as Macrophage Scavenger Receptor 1 (MSR 1), Scavenger Receptor A (SR-A) or SCARA1. CD204 belongs to the class A family of scavenger receptors that are primarily responsible for binding a wide variety of macromolecules and are involved in many aspects of macrophage activity. Other members of this family include: MARCO (SCARA2), CSR (SCARA3), SRCL (SCARA4) and SCARA5. All class A scavenger receptors are trans-membrane proteins with extracellular C-termini and homotrimeric structures. CD204 exists as 2 splice variants: the more ubiquitous full length isoform (type I) and a shorter isoform (type II) that lacks the C-terminal cysteine-rich domain. Both types are functional and bind modified LDL with similar affinity. In addition to lipid metabolism, CD204 plays an important role suppressing inflammation and macrophage activation. CD204 is considered one of the markers of M2 and Tumor-Associated Macrophages. Interestingly, this molecule is responsible for adherence of RAW264.7 cells to tissue culture-treated plastic in the absence of Ca and Mg cations. This M204PA antibody binds to CD204 of multiple mouse strains, including C57Bl/6, Balb/c and Swiss-Webster (outbred). This antibody works on fixed cells and can be used to stain CD204 intracellularly. The M204PA antibody does not cross-react with human or rat. This antibody is not suitable for IHC staining.
CD204 gene encodes the class A macrophage scavenger receptors, which include three different types (1, 2, 3) generated by alternative splicing of this gene. These receptors or isoforms are macrophage-specific trimeric integral membrane glycoproteins and have been implicated in many macrophage-associated physiological and pathological processes including atherosclerosis, Alzheimer's disease, and host defense. The isoforms type 1 and type 2 are functional receptors and are able to mediate the endocytosis of modified low density lipoproteins (LDLs). The isoform type 3 does not internalize modified LDL (acetyl-LDL) despite having the domain shown to mediate this function in the types 1 and 2 isoforms. It has an altered intracellular processing and is trapped within the endoplasmic reticulum, making it unable to perform endocytosis. The isoform type 3 can inhibit the function of isoforms type 1 and type 2 when co-expressed, indicating a dominant negative effect and suggesting a mechanism for regulation of scavenger receptor activity in macrophages. Diseases associated with MSR1 include Barrett Esophagus and Prostate Cancer.Spezifikation
CD204 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide | |
P30204 | |
MSR1 | |
Affinity chromatography | |
RUO | |
20288 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
M204PA | |
PE | |
MSR1 | |
bovine scavenger receptor type II; CD204; Macrophage acetylated LDL receptor I and II; macrophage scavenger receptor 1; macrophage scavenger receptor type III; macrophage scavenger receptor types I and II; MRS-A; MSR; Msr1; MSR-A; phSR1; phSR2; RGD1564316; SCARA1; Scavenger receptor class A member 1; scavenger receptor class A, member 1; scavenger receptor type A; scavenger receptor type I; Scvr; SRA; SR-A; SR-AI; SR-AI/II; SR-AII | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.