Learn More
CD23 Monoclonal Antibody (EBVCS2), Super Bright™ 600, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
151.00€ - 333.00€
Spécification
Antigène | CD23 |
---|---|
Clone | EBVCS2 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15973474
|
Affymetrix eBioscience
63-0238-41 |
25 Tests |
151.00€
25 pièces |
Expédition estimée: 24-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15983474
|
Affymetrix eBioscience
63-0238-42 |
100 Tests |
333.00€
100 pièces |
Expédition estimée: 16-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Description: The EBVCS2 monoclonal antibody reacts with human CD23, a 45 kDa type II transmembrane glycoprotein. CD23 is expressed on mature B cells, mantle zone B cells, follicular dendritic cells and at low levels on T, NK, langerhans cells and platelets. Expression of CD23 is upregulated upon B cell activation, and soluble forms of the antigen have been reported to be biologically active. CD23 is a low affinity receptor for IgE and is thought to play a role in the regulation of IgE response and B cell activation. CD21 and the alpha subunit of CD11b and CD11c bind to CD23. Applications Reported: This EBVCS2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This EBVCS2 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Super Bright 600 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 600 nm. We recommend using a 610/20 bandpass filter. Please make sure that your instrument is capable of detecting this fluorochrome. When using two or more Super Bright dye-conjugated antibodies in a staining panel, it is recommended to use Super Bright Staining Buffer (Product No.
CD23 Monoclonal antibody specifically detects CD23 in Human samples. It is validated for Flow Cytometry.Spécification
CD23 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
BLAST-2; CD23; CD23 antigen; CD23A; CLEC4J; C-type lectin domain family 4 member J; C-type lectin domain family 4, member J; Fc epsilon receptor II; FC epsilon RII; Fc fragment of IgE receptor II; Fc fragment of IgE, low affinity II, receptor for (CD23); Fc receptor alpha multiple ligand receptor; Fc receptor, IgE, low affinity II, alpha polypeptide; Fcalpha muR; Fcalpha RII; Fce2; fc-epsilon-RII; FCER2; Fcer2a; FceRII; IGEBF; Immunoglobulin E-binding factor; immunoglobulin epsilon-chain; low affinity immunoglobulin epsilon Fc receptor; Low affinity immunoglobulin epsilon Fc receptor membrane-bound form; Low affinity immunoglobulin epsilon Fc receptor soluble form; low-affinity IgE receptor; Ly-42; lymphocyte IgE receptor | |
FCER2 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
FCER2 |
EBVCS2 | |
Flow Cytometry | |
Super Bright 600 | |
Mouse | |
Human | |
P06734 | |
2208 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |