missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD268 (BAFF Receptor) Mouse anti-Human, Alexa Fluor 647, Clone: 11C1, BD Pharmingen™
Mouse Monoclonal Antibody
Marque: BD Biosciences 564817
Informations supplémentaires : Poids : 0.01000kg
Description
CD268 (BAFF Receptor) Monoclonal antibody specifically detects CD268 (BAFF Receptor) in Human samples. It is validated for Flow cytometry.
CD268 (BAFF Receptor) Monoclonal antibody specifically detects CD268 (BAFF Receptor) in Human samples. It is validated for Flow cytometry.Spécification
CD268 (BAFF Receptor) | |
Monoclonal | |
Alexa Fluor 647 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Cell Biology | |
Human | |
Purified |
Flow Cytometry | |
11C1 | |
Vol. Per test: 5μL | |
BAFF Receptor; BAFF-R; BLyS receptor 3; BR3; TNFRSF13C | |
Human BAFF-R Transfected Cell Line | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions and unreacted Alexa Fluor™ 647 was removed. | |
IgG1 κ |
For Research Use Only.