missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 epsilon Antibody (TR66) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 5 publications
Marque: Novus Biologicals NBP1-97446-0.025MG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD3 epsilon Monoclonal specifically detects CD3 epsilon in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Spécification
| CD3 epsilon | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1:10-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | |
| CD3e antigen, CD3e antigen, epsilon polypeptide (TiT3 complex), CD3e molecule, epsilon (CD3-TCR complex), CD3-epsilon, FLJ18683, T3E, T-cell antigen receptor complex, epsilon subunit of T3, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain, TCRE | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Recognizes the epsilon-chain of human CD3. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Immunofluorescence, Flow Cytometry, Immunocytochemistry, Immunoprecipitation | |
| TR66 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| CD3E | |
| Cloned human helper T cell line. | |
| 0.025 mg | |
| Adaptive Immunity, Apoptosis, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Lines, Stem Cell Markers | |
| 916 | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu