missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD3 gamma Mouse anti-Human, Clone: 2E9G7, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 60347-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
CD3G is the CD3-gamma polypeptide, which together with CD3-epsilon, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. Defects in the CD3G gene are associated with T cell immunodeficiency.Spécification
CD3 gamma | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P09693 | |
CD3G | |
CD3 gamma Fusion Protein Ag15502 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2E9G7 | |
Unconjugated | |
CD3G | |
CD3 gamma, CD3G, T cell receptor T3 gamma chain, T3G | |
Mouse | |
Protein A | |
RUO | |
917 | |
-20°C | |
Liquid |