missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, NovaFluor™ Yellow 660, Clone: UCHT1, Invitrogen™

Mouse Monoclonal Antibody

Marque:  Invitrogen H002T03Y04

Informations supplémentaires : Poids : 0.02000kg

 Afficher plus de versions de ce produit

Code produit. 17266416

  • 350.00€ / 100 pièces
Expédition estimée: 19-12-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The UCHT1 monoclonal antibody reacts with human CD3e, a 20 kDa subunit of the TCR complex. Along with the other CD3 subunits gamma and delta, the epsilon chain is required for proper assembly, trafficking and surface expression of the TCR complex. CD3 is expressed by thymocytes in a developmentally regulated manner and by all mature T cells. Crosslinking of TCR via immobilized UCHT1 initiates an intracellular biochemical pathway resulting in cellular activation and proliferation.
Applications Reported: The UCHT1 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This UCHT1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 4 μL (0.6 μg) per test. A test is defined as the amount ( μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
NovaFluor dyes are not compatible with DNA intercalating viability dyes. Do not use viability dyes such as propidium iodide, 7-actinomycin D (7-AAD) and DAPI. Invitrogen LIVE/DEAD Fixable Dead Cell stains are recommended for use with NovaFluor dyes.
When using more than one NovaFluor dye, CellBlox Monocyte and Macrophage Blocking Buffer may be used to block non-specific binding of dyes to monocytes.
Excitation: 552 nm; Emission: 663 nm; Laser: 561 nm (Yellow) Laser

CD3 Monoclonal antibody specifically detects CD3 in Human samples. It is validated for Flow Cytometry
Spécification

Spécification

CD3
Monoclonal
4 μL/Test
PBS with no preservative; pH 7.2
P04234, P07766, P09693, P20963
Mouse
100 Tests
Primary
Human
Antibody
IgG1 κ
Flow Cytometry
UCHT1
NovaFluor Yellow 660
Cd3d
4930549J05Rik; A430104F18Rik; AI504783; antigen CD3D, delta polypeptide (TiT3 complex); antigen CD3E, epsilon polypeptide (TiT3 complex); antigen CD3G, gamma polypeptide; antigen CD3Z, zeta polypeptide; AW552088; CD247; CD247 antigen; CD247 antigen, zeta subunit; CD247 molecule; Cd3; CD3 antigen delta chain; CD3 antigen delta polypeptide; CD3 antigen gamma chain; CD3 antigen, delta polypeptide; CD3 antigen, delta subunit; CD3 antigen, epsilon polypeptide; CD3 antigen, gamma polypeptide; CD3 antigen, zeta polypeptide; CD3 delta; CD3 epsilon; CD3 epsilon chain; CD3 epsilon subunit; CD3 epsilon subunit precursor; CD3 gamma-chain; CD3 glycoprotein; CD3 molecule delta polypeptide; CD3 molecule, delta; CD3 molecule, epsilon; CD3 molecule, epsilon polypeptide; CD3 molecule, gamma; CD3 molecule, gamma polypeptide; CD3 protein; CD3 type I transmembrane glycoprotein; CD3 zeta chain; Cd3d; CD3D antigen delta; CD3d antigen, delta polypeptide (TiT3 complex); CD3d molecule; CD3d molecule, delta (CD3-TCR complex); CD3-DELTA; CD3e; CD3E antigen, epsilon polypeptide; CD3E antigen, epsilon polypeptide (TiT3 complex); CD3e molecule; CD3e molecule, epsilon (CD3-TCR complex); CD3epsilon; CD3-epsilon; Cd3-eta; CD3g; CD3G antigen, gamma polypeptide; CD3g antigen, gamma polypeptide (TiT3 complex); CD3g molecule; CD3g molecule, epsilon (CD3-TCR complex); CD3g molecule, gamma (CD3-TCR complex); CD3-GAMMA; CD3H; CD3Q; CD3Z; CD3Z antigen, zeta polypeptide (TiT3 complex); Cd3zeta; CD3-ZETA; CD3zeta chain; CD3-zeta/eta; Ctg3; Ctg-3; FLJ18683; IMD17; IMD18; IMD19; IMD25; Leu-4; OKT3, delta chain; T3/TCR complex; T3D; T3E; T3G; T3z; T-cell antigen receptor complex, epsilon subunit of T3; T-cell antigen receptor complex, gamma subunit of T3; T-cell antigen receptor complex, zeta subunit of CD3; T-cell receptor CD3 epsilon chain; T-cell receptor CD3 epsilon subunit; T-cell receptor CD3 subunit zeta; T-cell receptor CD3, subunit zeta; T-cell receptor T3 delta chain; T-cell receptor T3 eta chain; T-cell receptor T3 gamma chain; T-cell receptor T3 zeta chain; T-cell receptor zeta chain; T-cell surface antigen T3/Leu-4 epsilon chain; T-cell surface glycoprotein CD3 delta chain; T-cell surface glycoprotein CD3 epsilon chain; T-cell surface glycoprotein CD3 gamma chain; T-cell surface glycoprotein CD3 zeta chain; T-cell surface protein; TcR CD3 delta-chain; TcR CD3 gamma-chain; TCR zeta chain; TCR zeta chain subunit; TCRE; Tcrk; TCRZ; TCRzeta; TiT3 complex
Affinity chromatography
RUO
915, 916, 917, 919
4° C, store in dark, DO NOT FREEZE!
Liquid
Vidéos
FDS
Documents
Certificats
Promotions

Promotions