Learn More
CD3 Mouse, anti-Human, R718, Clone: SP34-2, BD Horizon™
Mouse Monoclonal Antibody
Marque:  BD Biosciences 566956
Informations supplémentaires : Poids : 0.00020kg
Code produit. 17257744
  • 161.00€ / 25 tests
Expédition estimée: 13-10-2022
Description
Clone SP34-2 is a mouse IgG1 isotype monoclonal antibody, descendant of SP34 (mouse IgG3), with the same specificity and reactivity pattern as the parent clone. It cross-reacts with a major subset of peripheral blood lymphocytes, but not monocytes or granulocytes, of baboon, and rhesus, cynomolgus, and pigtail macaque monkeys. The distribution on lymphocytes is similar to that observed with normal human donor lymphocytes with the majority of CD3-positive cells being negative when dual stained with antibodies to B or NK cells markers. SP34-2 is also capable of inducing cell proliferation on both human and non-human primate PBMC.
Spécification
Spécification
CD3
Monoclonal
R718
25 Tests
Primary
Refrigerate Upon Arrival
Flow Cytometry
SP34-2