missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD300LG/Nepmucin Antibody, Janelia Fluor™ 549, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-98158JF549
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
CD300LG/Nepmucin Polyclonal antibody specifically detects CD300LG/Nepmucin in Rat samples. It is validated for ELISA
Spécification
| CD300LG/Nepmucin | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 146894 | |
| Purified |
| Polyclonal | |
| ELISA | |
| CD300 antigen like family member G, CD300 antigen-like family member G, CD300 molecule like family member g, CD300 molecule-like family member g, CD300g antigen, CLM-9, CLM9Trem4, CMRF35-like molecule 9, NEPMUCIN, TREM4, TREM-4, Triggering receptor expressed on myeloid cells 4 | |
| Produced in rabbits immunized with purified, recombinant Rat CD300LG/Nepmucin (Accession#: XP_002724611.1; Met1-Arg160). | |
| 0.1 ml | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu